1-Boc-6-methyl-2-indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
6-Methyl-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-methyl-1H-indol-2-yl)boronic acid
PN-2084-5G
PN-2084-1G
COBBPN-2084-5GEA
1590
EUR
COBBPN-2084-5G
COBBPN-2084-1G
1-Boc-6-methyl-2-indolboronsäurepinakolester
1-Boc-6-methyl-2-indolboronsäurepinakolester
Formel:
C₁₄H₁₈BNO₄ Molecular Weight: 275,11 g/mol |
MDL:
MFCD04973738 CAS-Nummer: 850568-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...