1-Boc-6-methyl-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methyl-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-methyl-1H-indol-2-yl)boronic acid

BB-2084-5G BB-2084-1G
COBBBB-2084-5GEA 1250 EUR
COBBBB-2084-5G COBBBB-2084-1G
1-Boc-6-methyl-2-indolboronsäurepinakolester
1-Boc-6-methyl-2-indolboronsäurepinakolester
Formel: C₁₄H₁₈BNO₄
Molecular Weight: 275.11 g/mol
MDL: MFCD04973738
CAS-Nummer: 850568-51-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR