1-Butyl-3-methylimidazoliumacetat, Sigma-Aldrich®

Lieferant: Merck

39952-1KG-F 39952-100G-F
SIAL39952-1KG-FEA 1770 EUR
SIAL39952-1KG-F SIAL39952-100G-F
1-Butyl-3-methylimidazoliumacetat, Sigma-Aldrich®
1-Butyl-3-methylimidazolium acetate
Formel: C₁₀H₁₈N₂O₂
Molecular Weight: 198.27 g/mol
MDL: MFCD06798175
CAS-Nummer: 284049-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR