1-Butyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Butyl-3-methylimidazolium Bis(trifluoromethanesulphonyl)imide, 1-Butyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide

711713-100G 711713-1KG
SIAL711713-100GEA 329 EUR
SIAL711713-100G SIAL711713-1KG
1-Butyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid
Formel: C₁₀H₁₅F₆N₃O₄S₂
Molecular Weight: 419,37 g/mol
MDL: MFCD05664714
CAS-Nummer: 174899-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR