1-Butyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Butyl-3-methylimidazolium Bis(trifluoromethanesulphonyl)imide, 1-Butyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide
77896-1G-F
77896-5G-F
SIAL77896-1G-FEA
152
EUR
SIAL77896-1G-F
SIAL77896-5G-F
1-Butyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid
Formel:
C₁₀H₁₅F₆N₃O₄S₂ Molecular Weight: 419.37 g/mol |
MDL:
MFCD05664714 CAS-Nummer: 174899-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...