Phloroglucintrimethylether ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
O,O,O-1,3,5-Trimethylresorcin, 1,3,5-Trimethoxybenzol
A13981.09
A13981.18
A13981.30
A13981.09EA
26.8
EUR
A13981.09
A13981.18
A13981.30
Phloroglucintrimethylether ≥99%
Phloroglucintrimethylether
Purity: 99%
CAS No.: 621-23-8
Molecular Formula: C9H12O3
Formula Weight: 168.19
Flash Point: 85°(185°F)
Melting Point: 51-53°
Boiling Point: 255°
Beilstein Reference: 1307993
MDL No.: MFCD00008385
Formel:
C₉H₁₂O₃ Molecular Weight: 168,19 g/mol Siedepunkt: 255 °C Schmelzpunkt: 51…53 °C Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008385 CAS-Nummer: 621-23-8 EINECS: 210-673-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...