2-(Brommethyl)-2-(hydroxymethyl)-1,3-propandiol

Lieferant: CHEMPUR

Synonyme: 2-(Bromomethyl)-2-(hydroxymethyl)propane-1,3-diol

RJC01041.250 RJC01041.1
CHMPRJC01041.250EA 0 EUR
CHMPRJC01041.250 MAYBRJC01041.1
2-(Brommethyl)-2-(hydroxymethyl)-1,3-propandiol
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol
Formel: C₅H₁₁BrO₃
Molecular Weight: 199,04 g/mol
MDL: MFCD00135306
CAS-Nummer: 19184-65-7
EINECS: 242-863-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR