3-Brom-5-nitrobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

702986-1G 702986-250MG
SIAL702986-1GEA 104 EUR
SIAL702986-1G SIAL702986-250MG
3-Brom-5-nitrobenzaldehyd, Sigma-Aldrich®
3-Bromo-5-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
Molecular Weight: 230,02 g/mol
MDL: MFCD00100097
CAS-Nummer: 355134-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR