4-Chlor-3-sulfamoylbenzoesäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
293598-1G
293598-5G
SIAL293598-1GEA
53.9
EUR
SIAL293598-1G
SIAL293598-5G
4-Chlor-3-sulfamoylbenzoesäure, Sigma-Aldrich®
4-Chlor-3-sulfamoylbenzoesäure
Formel:
C₇H₆ClNO₄S Molecular Weight: 235.65 g/mol |
MDL:
MFCD00012375 CAS-Nummer: 1205-30-7 EINECS: 214-882-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...