4-Sulfamoylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Sulphamoylbenzoic acid

M-Clarity™ Qualitätsstufe = MQ200

C11804-100G C11804-5G
SIALC11804-100GEA 181 EUR
SIALC11804-100G SIALC11804-5G
4-Sulfamoylbenzoesäure, Sigma-Aldrich®
4-Sulfamoylbenzoesäure
Formel: C₇H₇NO₄S
Molecular Weight: 201.2 g/mol
MDL: MFCD00007938
CAS-Nummer: 138-41-0
EINECS: 205-327-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR