Diethylethoxymethylenmalonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Diethyl 2-(ethoxymethylene)malonate
M-Clarity™ Qualitätsstufe = MQ100
D94208-100G
D94208-500G
SIALD94208-100GEA
49.8
EUR
SIALD94208-100G
SIALD94208-500G
Diethylethoxymethylenmalonat, Sigma-Aldrich®
Diethylethoxymethylenmalonat
Formel:
C₁₀H₁₆O₅ Molecular Weight: 216,23 g/mol |
MDL:
MFCD00009148 CAS-Nummer: 87-13-8 EINECS: 201-725-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...