5-Benzyloxytryptamin Hydrochlorid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
106110010
106112500
106110050
ACRO106110010EA
153
EUR
ACRO106110010
ACRO106112500
ACRO106110050
5-Benzyloxytryptamin Hydrochlorid 98%
5-Benzyloxytryptamin Hydrochlorid
Formel:
C₁₇H₁₉ClN₂O Schmelzpunkt: 255…259 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00012685 CAS-Nummer: 52055-23-9 EINECS: 257-626-4 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
HPLC | ≥97.5 % |
Stabilizer | 20 to 60 ppm amylene |
Residue after evaporation | ≤0.002 % |
Free halogens | passes test |
Titrable acid | ≤0.0003 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...