5-Benzyloxytryptamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-[5-(Benzyloxy)-1H-indol-3-yl]ethanamine hydrochloride, 5-(phenylmethoxy)indole-3-ethanamine monohydrochloride

M-Clarity™ Qualitätsstufe = MQ100

B28656-1G B28656-250MG
SIALB28656-1GEA 134 EUR
SIALB28656-1G SIALB28656-250MG
5-Benzyloxytryptamin Hydrochlorid, Sigma-Aldrich®
5-Benzyloxytryptamin Hydrochlorid
Formel: C₁₇H₁₉ClN₂O
MDL: MFCD00012685
CAS-Nummer: 52055-23-9
EINECS: 257-626-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR