2,3-Dichlorbenzaldehydoxim

Lieferant: Thermo Scientific

Synonyme: 2,3-Dichlorobenzaldoxime

SPB01023.1 SPB01023.10 SPB01023.50
MAYBSPB01023.1EA 0 EUR
MAYBSPB01023.1 MAYBSPB01023.10 MAYBSPB01023.50
2,3-Dichlorbenzaldehydoxim
2,3-Dichlorbenzaldehydoxim
Formel: C₇H₅Cl₂NO
Molecular Weight: 190,03 g/mol
CAS-Nummer: 4414-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR