2,4-Dichlorbenzaldehydoxim
Lieferant: Thermo Scientific
SPB04865DA
SPB04865EA
SPB04865.50
MAYBSPB04865DAEA
0
EUR
MAYBSPB04865DA
MAYBSPB04865EA
MAYBSPB04865.50
2,4-Dichlorbenzaldehydoxim
2,4-Dichlorbenzaldehydoxim
Formel:
C₇H₅Cl₂NO |
CAS-Nummer:
56843-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...