2-Chlorbenzaldehydoxim

Lieferant: MENAI

E2651.50 E2651.100
MENAE2651.50EA 0 EUR
MENAE2651.50 MENAE2651.100
2-Chlorbenzaldehydoxim
2-Chlorbenzaldehydoxim
Formel: C₇H₆ClNO
Molecular Weight: 155.58
MDL: MFCD00016406
CAS-Nummer: 3717-28-0
EINECS: 223-065-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR