4-Fluorbenzaldehydoxim

Lieferant: FLUOROCHEM

011169-1G 011169-10G
FLUO011169-1GEA 31.1 EUR
FLUO011169-1G FLUO011169-10G
4-Fluorbenzaldehydoxim
4-Fluorbenzaldehydoxim
Formel: C₇H₆FNO
MDL: MFCD00133101
CAS-Nummer: 459-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR