4-Fluorbenzaldehydoxim

Lieferant: MENAI

DH570.10 DH570.1
MENADH570.10EA 0 EUR
MENADH570.10 MENADH570.1
4-Fluorbenzaldehydoxim
4-Fluorbenzaldehydoxim
Formel: C₇H₆FNO
Molecular Weight: 139.13
MDL: MFCD00133101
CAS-Nummer: 459-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR