o-Dianisidin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3'-Dimethoxybenzidine dihydrochloride

M-Clarity™ Qualitätsstufe = MQ200

D9154-100TAB D9154-50TAB
SIALD9154-100TABEA 378 EUR
SIALD9154-100TAB SIALD9154-50TAB
o-Dianisidin Dihydrochlorid, Sigma-Aldrich®
o-Dianisidin Dihydrochlorid
Formel: C₁₄H₁₈Cl₂N₂O₂
Molecular Weight: 317,21 g/mol
MDL: MFCD00012488
CAS-Nummer: 20325-40-0
EINECS: 243-737-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR