Nerol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: cis-3,7-Dimethyl-2,6-octadien-1-ol

376540250 376541000 376545000
ACRO376540250EA 27.5 EUR
ACRO376540250 ACRO376541000 ACRO376545000
Nerol 97%
Nerol
Formel: C₁₀H₁₈O
Molecular Weight: 154,25 g/mol
Flammpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063204
CAS-Nummer: 106-25-2
EINECS: 203-378-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4790 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR