Geraniol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (E)-3,7-Dimethylocta-2,6-dien-1-ol

M-Clarity™ Qualitätsstufe = MQ300

W250716-1KG-K W250716-4KG-K W250716-9KG-K
SIALW250716-1KG-KEA 217 EUR
SIALW250716-1KG-K SIALW250716-4KG-K SIALW250716-9KG-K
Geraniol, Sigma-Aldrich®
Geraniol
Formel: C₁₀H₁₈O
Molecular Weight: 154.25 g/mol
MDL: MFCD00002917
CAS-Nummer: 106-24-1
EINECS: 203-377-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR