1-Boc-2-piperidon ≥99%

Lieferant: Thermo Fisher Scientific

L18896.03 L18896.06
L18896.03EA 13.1 EUR
L18896.03 L18896.06
1-Boc-2-piperidon ≥99%
1-Boc-2-piperidon

Purity: 99% CAS No.: 85908-96-9 Molecular Formula: C10H17NO3 Formula Weight: 199.25 Melting Point: 34-37° Beilstein Reference: 4674955

Formel: C₁₀H₁₇NO₃
Schmelzpunkt: 34…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179046
CAS-Nummer: 85908-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR