1-Boc-3-piperidon ≥97%
Lieferant: Thermo Scientific
Synonyme:
N-BOC-3-piperidone, 1-Boc-3-Piperidinone
L19276.03
L19276.06
L19276.03EA
29
EUR
L19276.03
L19276.06
1-Boc-3-piperidon ≥97%
1-Boc-3-piperidon
Purity: 97%
CAS No.: 98977-36-7
Molecular Formula: C10H17NO3
Formula Weight: 199.25
Melting Point: 37-39°
Beilstein Reference: 5936353
Formel:
C₁₀H₁₇NO₃ Molecular Weight: 199,25 g/mol Schmelzpunkt: 37…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631193 CAS-Nummer: 98977-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...