1-Boc-4-piperidon

Lieferant: FLUOROCHEM

Synonyme: 4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone

019201-25G 019201-5G
FLUO019201-25GEA 30.8 EUR
FLUO019201-25G FLUO019201-5G
1-Boc-4-piperidon
1-Boc-4-piperidon
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,25 g/mol
MDL: MFCD00151800
CAS-Nummer: 79099-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR