(R)-(+)-4-Brom-α-methylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-1-(4-Bromophenyl)ethanol

M-Clarity™ Qualitätsstufe = MQ100

686336-1G 686336-250MG
SIAL686336-1GEA 164 EUR
SIAL686336-1G SIAL686336-250MG
(R)-(+)-4-Brom-α-methylbenzylalkohol, Sigma-Aldrich®
(R)-(+)-4-Brom-α-methylbenzylalkohol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD06201861
CAS-Nummer: 76155-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR