(S)-(-)-4-Fluor-α-methylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-1-(4-Fluorophenyl)ethanol, (1S)-1-(4-Fluorophenyl)ethan-1-ol

M-Clarity™ Qualitätsstufe = MQ100

685852-1G 685852-250MG
SIAL685852-1GEA 208 EUR
SIAL685852-1G SIAL685852-250MG
(S)-(-)-4-Fluor-α-methylbenzylalkohol, Sigma-Aldrich®
(S)-(-)-4-Fluor-α-methylbenzylalkohol
Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
MDL: MFCD03092998
CAS-Nummer: 101219-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR