1-Boc-piperazin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

AM-1697-100G AM-1697-25G
COBBAM-1697-100GEA 101 EUR
COBBAM-1697-100G COBBAM-1697-25G
1-Boc-piperazin
1-Boc-piperazin
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186,25 g/mol
MDL: MFCD00075265
CAS-Nummer: 57260-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR