Trimethylsilylisocyanat

Lieferant: CHEMPUR SILANE

SILS19575.5 SILS19575.25
SILNSILS19575.5EA 0 EUR
SILNSILS19575.5 SILNSILS19575.25
Trimethylsilylisocyanat
Trimethylsilylisocyanat
Formel: C₄H₉NOSi
MDL: MFCD00001993
CAS-Nummer: 1118-02-1
EINECS: 214-256-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR