1-Benzyl-4-piperidon

Lieferant: FLUOROCHEM

Synonyme: N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone

023286-100G 023286-25G
FLUO023286-100GEA 82.1 EUR
FLUO023286-100G FLUO023286-25G
1-Benzyl-4-piperidon
1-Benzyl-4-piperidon
Formel: C₁₂H₁₅NO
Molecular Weight: 189,26 g/mol
MDL: MFCD00006192
CAS-Nummer: 3612-20-2
EINECS: 222-782-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR