4-Bromoxindol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromooxindole, 4-Bromo-2,3-dihydro-1H-indol-2-one, 4-Bromoindolin-2-one
IN-0117-5G
IN-0117-1G
COBBIN-0117-5GEA
221
EUR
COBBIN-0117-5G
COBBIN-0117-1G
4-Bromoxindol
4-Bromoxindol
Formel:
C₈H₆BrNO Molecular Weight: 212,05 g/mol |
MDL:
MFCD06659911 CAS-Nummer: 99365-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...