5-Bromoxindol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-1,3-dihydro-2H-indol-2-one, 5-Bromoindolin-2-one, 5-Bromo-2-oxindole

H50573.03 H50573.06
H50573.03EA 88.2 EUR
H50573.03 H50573.06
5-Bromoxindol ≥98%
5-Bromoxindol

Purity: 98% CAS No.: 20870-78-4 Molecular Formula: C8H6BrNO Formula Weight: 212.05 MDL No.: MFCD00456998

Formel: C₈H₆BrNO
Molecular Weight: 212,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00456998
CAS-Nummer: 20870-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR