6-Bromoxindol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromo-2-oxindole, 6-Bromoindolin-2-one

IN-0081-5G IN-0081-1G
COBBIN-0081-5GEA 63.4 EUR
COBBIN-0081-5G COBBIN-0081-1G
6-Bromoxindol
6-Bromoxindol
Formel: C₈H₆BrNO
Molecular Weight: 212.05 g/mol
MDL: MFCD02179605
CAS-Nummer: 99365-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR