6-Chloroxindol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Chloro-2-oxindole

IN-0102-5G IN-0102-100G IN-0102-25G
COBBIN-0102-5GEA 26.2 EUR
COBBIN-0102-5G COBBIN-0102-100G COBBIN-0102-25G
6-Chloroxindol
6-Chloroxindol
Formel: C₈H₆ClNO
Molecular Weight: 167,59 g/mol
MDL: MFCD00209962
CAS-Nummer: 56341-37-8
EINECS: 421-330-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR