6-Chloroxindol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Chloro-2-oxindole
M-Clarity™ Qualitätsstufe = MQ100
636215-25G
636215-5G
SIAL636215-25GEA
44.4
EUR
SIAL636215-25G
SIAL636215-5G
6-Chloroxindol, Sigma-Aldrich®
6-Chloroxindol
Formel:
C₈H₆ClNO Molecular Weight: 167,59 g/mol |
MDL:
MFCD00209962 CAS-Nummer: 56341-37-8 EINECS: 421-330-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...