5-Fluoroxindol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoroindolin-2-one, 5-Fluoro-2-oxoindoline, 5-Fluoro-2-oxindole

IN-0072-1G IN-0072-5G
COBBIN-0072-1GEA 0 EUR
COBBIN-0072-1G COBBIN-0072-5G
5-Fluoroxindol
5-Fluoroxindol
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
MDL: MFCD02179598
CAS-Nummer: 56341-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR