6-Fluoroxindol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Fluoroindolin-2-one

IN-0086-1G IN-0086-25G IN-0086-5G
COBBIN-0086-1GEA 76.1 EUR
COBBIN-0086-1G COBBIN-0086-25G COBBIN-0086-5G
6-Fluoroxindol
6-Fluoroxindol
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
MDL: MFCD00047212
CAS-Nummer: 56341-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR