3-Methyloxindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methyl-2-oxindole

M-Clarity™ Qualitätsstufe = MQ100

493937-25G 493937-5G
SIAL493937-25GEA 291 EUR
SIAL493937-25G SIAL493937-5G
3-Methyloxindol, Sigma-Aldrich®
3-Methyloxindol
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD04037370
CAS-Nummer: 1504-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR