1-Boc-5-bromindol
Lieferant: COMBI-BLOCKS
Synonyme:
N-Boc-5-bromoindole, 5-Bromo-1-tert-butoxycarbonylindole, tert-Butyl 5-bromo-1H-indole-1-carboxylate
IN-0233-5G
IN-0233-25G
IN-0233-1G
COBBIN-0233-5GEA
59.3
EUR
COBBIN-0233-5G
COBBIN-0233-25G
COBBIN-0233-1G
1-Boc-5-bromindol
1-Boc-5-bromindol
Formel:
C₁₃H₁₄BrNO₂ Molecular Weight: 296,16 g/mol |
MDL:
MFCD02090067 CAS-Nummer: 182344-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...