1-Boc-6-bromindol

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 6-bromo-1H-indole-1-carboxylate

IN-0225-1G IN-0225-5G
COBBIN-0225-1GEA 127 EUR
COBBIN-0225-1G COBBIN-0225-5G
1-Boc-6-bromindol
1-Boc-6-bromindol
Formel: C₁₃H₁₄BrNO₂
Molecular Weight: 296.16 g/mol
MDL: MFCD09954826
CAS-Nummer: 147621-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR