1-Boc-4-chlorindol

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 4-chloro-1H-indole-1-carboxylate

IN-0227-5G IN-0227-25G IN-0227-1G
COBBIN-0227-5GEA 303 EUR
COBBIN-0227-5G COBBIN-0227-25G COBBIN-0227-1G
1-Boc-4-chlorindol
1-Boc-4-chlorindol
Formel: C₁₃H₁₄ClNO₂
Molecular Weight: 251.71 g/mol
MDL: MFCD09025758
CAS-Nummer: 129822-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR