1-Boc-5-chlorindol

Lieferant: COMBI-BLOCKS

Synonyme: 1-(tert-Butoxycarbonyl)-5-chloroindole, tert-Butyl 5-chloro-1H-indole-1-carboxylate

IN-0228-25G IN-0228-1G IN-0228-5G
COBBIN-0228-25GEA 791 EUR
COBBIN-0228-25G COBBIN-0228-1G COBBIN-0228-5G
1-Boc-5-chlorindol
1-Boc-5-chlorindol
Formel: C₁₃H₁₄ClNO₂
Molecular Weight: 251.71 g/mol
MDL: MFCD05865201
CAS-Nummer: 129822-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR