1-Boc-5-cyanindol
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 5-cyano-1H-indole-1-carboxylate
IN-0129-5G
IN-0129-25G
IN-0129-1G
COBBIN-0129-5GEA
102
EUR
COBBIN-0129-5G
COBBIN-0129-25G
COBBIN-0129-1G
1-Boc-5-cyanindol
1-Boc-5-cyanindol
Formel:
C₁₄H₁₄N₂O₂ Molecular Weight: 242,28 g/mol |
MDL:
MFCD08056374 CAS-Nummer: 475102-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...