3-Acetoxyindol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate
M-Clarity™ Qualitätsstufe = MQ200
8.20706.0001
8.20706.0005
8.20706.0001EA
67.3
EUR
8.20706.0001
8.20706.0005
3-Acetoxyindol, Sigma-Aldrich®
3-Acetoxyindol
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol |
MDL:
MFCD00014561 CAS-Nummer: 608-08-2 EINECS: 210-154-2 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 127 - 129 °C |
Molar Mass | 175.19 g/mol |
Synonyms | Acetic acid 3-indolyl ester, 3-Acetoxyindole, Indoxyl acetate |
Hill Formula | C10H9NO2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...