3-Acetoxyindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate

M-Clarity™ Qualitätsstufe = MQ200

I3500-1G I3500-250MG I3500-5G
SIALI3500-1GEA 111 EUR
SIALI3500-1G SIALI3500-250MG SIALI3500-5G
3-Acetoxyindol, Sigma-Aldrich®
3-Acetoxyindol
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00014561
CAS-Nummer: 608-08-2
EINECS: 210-154-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR