3-Acetoxyindol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Indolyl Acetate, Indoxyl acetate, 3-Indoxyl acetate, 3-Indolyl acetate

156660010 156660050
ACRO156660010EA 56.2 EUR
ACRO156660010 ACRO156660050
3-Acetoxyindol 97%
3-Acetoxyindol
Formel: C₁₀H₉NO₂
Molecular Weight: 175.19 g/mol
Schmelzpunkt: 128…131 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00014561
CAS-Nummer: 608-08-2
EINECS: 210-154-2

Bestellen

Spezifikation Testergebnisse

Appearance White to pink Powder or crystals
Infrared spectrum Conforms
Melting point 127 to 131 °C
GC ≥96.0 %
UV at 500 nm A: ≤0.01
pH 6.5 to 8.5
Titration with HCl ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR