4-Acetoxyindol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Indolyl acetate

IN-0029-1G IN-0029-5G
COBBIN-0029-1GEA 88.6 EUR
COBBIN-0029-1G COBBIN-0029-5G
4-Acetoxyindol
4-Acetoxyindol
Formel: C₁₀H₉NO₂
Molecular Weight: 175.19 g/mol
MDL: MFCD00010678
CAS-Nummer: 5585-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR