Methyl-3-indolcarboxylat ≥99%

Lieferant: Thermo Scientific
Achtung

AC30928.1 AC30928EA AC30928.50
MAYBAC30928.1EA 0 EUR
MAYBAC30928.1 MAYBAC30928EA MAYBAC30928.50
Methyl-3-indolcarboxylat ≥99%
Methyl-3-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00189407
CAS-Nummer: 942-24-5

Bestellen

Spezifikation Testergebnisse

Appearance Light brown Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR