Methyl-3-indolcarboxylat ≥99%

Lieferant: Thermo Scientific

L14516.14 L14516.22
L14516.14EA 45.8 EUR
L14516.14 L14516.22
Methyl-3-indolcarboxylat ≥99%
Methyl-3-indolcarboxylat

Purity: 99% CAS No.: 942-24-5 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 149-152° Beilstein Reference: 142023

Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00189407
CAS-Nummer: 942-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR