Methyl-4-indolcarboxylat ≥99%
Lieferant: Thermo Scientific
L16021.03 L16021.06 AB180081.0001
L16021.03EA 30.4 EUR
L16021.03 L16021.06 ABCRAB180081.0001
Methyl-4-indolcarboxylat ≥99%
Methyl-4-indolcarboxylat
Purity: 99% CAS No.: 39830-66-5 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 68-71° Beilstein Reference: 141826
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191222
CAS-Nummer: 39830-66-5