Methyl-5-indolcarboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 1H-indole-5-carboxylate
IN-0008-5G
IN-0008-25G
COBBIN-0008-5GEA
63.9
EUR
COBBIN-0008-5G
COBBIN-0008-25G
Methyl-5-indolcarboxylat
Methyl-5-indolcarboxylat
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol |
MDL:
MFCD00153023 CAS-Nummer: 1011-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...